Anti-CD63 Mouse Monoclonal Antibody (FITC (Fluorescein))
Supplier: Biorbyt
Certificates
About this item
Type: Primary
Antigen: CD63 (CD63 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- BIRBORB12113-50
- BIRBORB12113-100
- Antigen symbol:
- CD63
- CD63
- Antigen name:
- CD63 molecule
- CD63 molecule
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Storage buffer:
- Phosphate-buffered solution 0,09% sodium azide and 0,2% (w/v) BSA
- Phosphate-buffered solution 0,09% sodium azide and 0,2% (w/v) BSA
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together