Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-LTB4-R1 Rabbit Polyclonal Antibody
 :  Biorbyt
 
undefined
Anti-LTB4-R1 Rabbit Polyclonal Antibody
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB6335-20
  • BIRBORB6335-5
  • BIRBORB6335-200
  • BIRBORB6335-100
  • Antigen symbol:
  • LTB4-R1
  • LTB4-R1
  • LTB4-R1
  • LTB4-R1
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • KLH conjugated synthetic peptide derived from human LTB4-R1 C-terminus.
  • KLH conjugated synthetic peptide derived from human LTB4-R1 C-terminus.
  • KLH conjugated synthetic peptide derived from human LTB4-R1 C-terminus.
  • KLH conjugated synthetic peptide derived from human LTB4-R1 C-terminus.
  • Form:
  • liquid
  • liquid
  • liquid
  • liquid
  • Molecular weight:
  • 39 kDa
  • 39 kDa
  • 39 kDa
  • 39 kDa
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Concentration:
  • 0,5 mg/ml
  • 0,5 mg/ml
  • 0,5 mg/ml
  • 0,5 mg/ml
  • Storage buffer:
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice
  • Ice

 

Frequently Bought Together