Anti-MBD3L1 Mouse Monoclonal Antibody [clone: 1C7]
Catalog # ABNOH00085509-M08
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Methyl-CpG-binding Domain protein 3-like 1
- Antigen symbol:MBD3L1
- Clonality:Monoclonal
- Clone:1C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:85509
- Antigen synonyms:MBD3L
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MBD3L1
Clonality: Monoclonal
Clone: 1C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human