Anti-NGB Mouse Monoclonal Antibody [clone: 1A11-A9]
Catalog # ABNOH00058157-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Neuroglobin
- Antigen symbol:NGB
- Clonality:Monoclonal
- Clone:1A11-A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:58157
- Antigen synonyms:Neuroglobin|Ngb
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: NGB
Clonality: Monoclonal
Clone: 1A11-A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human