Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-PSMD14 Mouse Monoclonal Antibody [clone: 4A10-E8]
Anti-PSMD14 Mouse Monoclonal Antibody [clone: 4A10-E8]
Catalog # ABNOH00010213-M01
Supplier:  Abnova
undefined
Anti-PSMD14 Mouse Monoclonal Antibody [clone: 4A10-E8]
Catalog # ABNOH00010213-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proteasome (prosome macropain) 26S Subunit non-ATPase 14
  • Antigen symbol:
    PSMD14
  • Clonality:
    Monoclonal
  • Clone:
    4A10-E8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    10213
  • Antigen synonyms:
    PAD1|RPN11|POH1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: PSMD14
Clonality: Monoclonal
Clone: 4A10-E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human