Anti-ZNF213 Mouse Monoclonal Antibody [clone: 3D3]
ABNOH00007760-M02
: Abnova
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 213
- Antigen symbol:ZNF213
- Clonality:Monoclonal
- Clone:3D3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7760
- Antigen synonyms:ZSCAN53|ZKSCAN21|CR53
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ZNF213
Clonality: Monoclonal
Clone: 3D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human