Anti-TCP1 Mouse Monoclonal Antibody [clone: 2B2-D6]
Catalog # ABNOH00006950-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:T-Complex 1
- Antigen symbol:TCP1
- Clonality:Monoclonal
- Clone:2B2-D6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6950
- Antigen synonyms:CCTa|T-complex protein 1|alpha subunit|D6S230E|TCP1|tailless complex polypeptide 1|t-complex 1|CCT-alpha|CCT1|TCP-1-alpha
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TCP1
Clonality: Monoclonal
Clone: 2B2-D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human