Anti-S100 A9 Rabbit Polyclonal Antibody
Catalog # ABNOH00006280-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:S100 calcium binding protein A9
- Antigen symbol:S100A9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6280
- Antigen synonyms:S100A9|S100 Calcium binding protein A9|CAGB|MRP14|CFAG|S100 calcium-binding protein A9|NIF|MAC387|MIF|L1AG|calgranulin B|60B8AG|CGLB|P14|S100 calcium-binding protein A9 (calgranulin B)|LIAG|S100 Calcium binding protein A9 (calgranulin B)
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: S100 A9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human