Anti-S100 A8 Mouse Polyclonal Antibody
Catalog # ABNOH00006279-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:S100 calcium binding protein A8
- Antigen symbol:S100A8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6279
- Antigen synonyms:L1Ag|CP-10|CFAG|CAGA|MRP8|NIF|MIF|CGLA|60B8AG|MA387|P8
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: S100 A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human