Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-NME2 Mouse Monoclonal Antibody [clone: 1D3]
Catalog # ABNOH00004831-M06
Supplier:  Abnova
undefined
Anti-NME2 Mouse Monoclonal Antibody [clone: 1D3]
Catalog # ABNOH00004831-M06
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nucleoside diphosphate kinase B
  • Antigen symbol:
    NME2
  • Clonality:
    Monoclonal
  • Clone:
    1D3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    4831
  • Antigen synonyms:
    NDP kinase B|C-myc purine-binding transcription factor PUF|nm23-H2|NDK B|Nucleoside diphosphate kinase B
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: NME2
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat