Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-MYD88 Rabbit Polyclonal Antibody
Anti-MYD88 Rabbit Polyclonal Antibody
Catalog # ABNOH00004615-D01P
Supplier:  Abnova
undefined
Anti-MYD88 Rabbit Polyclonal Antibody
Catalog # ABNOH00004615-D01P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Myeloid differentiation primary response 88
  • Antigen symbol:
    MYD88
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    4615
  • Antigen synonyms:
    MYD88D|myeloid differentiation primary response 88
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: MYD88
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat