Anti-MYD88 Rabbit Polyclonal Antibody
ABNOH00004615-D01
: Abnova
- Antibody type:Primary
- Antigen name:Myeloid differentiation primary response 88
- Antigen symbol:MYD88
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4615
- Antigen synonyms:MYD88D|myeloid differentiation primary response 88
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: MYD88
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat