Anti-MYD88 Mouse Polyclonal Antibody
Catalog # ABNOH00004615-B02P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Myeloid differentiation primary response 88
- Antigen symbol:MYD88
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4615
- Antigen synonyms:MYD88D|myeloid differentiation primary response 88
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: MYD88
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human