Anti-MBD1 Mouse Monoclonal Antibody [clone: 2C7]
ABNOH00004152-M02
: Abnova
- Antibody type:Primary
- Antigen name:Methyl-CpG binding domain protein 1
- Antigen symbol:MBD1
- Clonality:Monoclonal
- Clone:2C7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4152
- Antigen synonyms:CXXC3|PCM1|RFT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: MBD1
Clonality: Monoclonal
Clone: 2C7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human