Anti-GCSH Mouse Monoclonal Antibody [clone: 3D8-A12]
ABNOH00002653-M01
: Abnova
- Antibody type:Primary
- Antigen name:Glycine cleavage system protein H (aminomethyl carrier)
- Antigen symbol:GCSH
- Clonality:Monoclonal
- Clone:3D8-A12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2653
- Antigen synonyms:NKH|GCE
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: GCSH
Clonality: Monoclonal
Clone: 3D8-A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human