Anti-E2F6 Mouse Monoclonal Antibody [clone: 2B6-G9]
Catalog # ABNOH00001876-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:E2F transcription Factor 6
- Antigen symbol:E2F6
- Clonality:Monoclonal
- Clone:2B6-G9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1876
- Antigen synonyms:MGC111545|E2F6_HUMAN.|E2F binding site modulating activity protein|Transcription factor E2F6|E2F transcription factor 6 isoform 1|EMA|E2F transcription factor 6|E2F 6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: E2F6
Clonality: Monoclonal
Clone: 2B6-G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human