Anti-CHRNA4 Mouse Monoclonal Antibody [clone: 4E3]
Catalog # ABNOH00001137-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cholinergic receptor, nicotinic, alpha 4 (neuronal)
- Antigen symbol:CHRNA4
- Clonality:Monoclonal
- Clone:4E3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1137
- Antigen synonyms:nicotinic|BFNC|NACRA4|EBN|neuronal acetylcholine receptor alpha 4 subunit|alpha polypeptide 4|alpha 4 subunit|EBN1|cholinergic receptor|alpha 4|neuronal nicotinic acetylcholine receptor alpha-4 subunit|CHRNA4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CHRNA4
Clonality: Monoclonal
Clone: 4E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human