Anti-CAMK2B Mouse Monoclonal Antibody [clone: 2D7-D8]
Catalog # ABNOH00000816-M02A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Calcium/calmodulin-dependent Protein Kinase II Beta
- Antigen symbol:CAMK2B
- Clonality:Monoclonal
- Clone:2D7-D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:816
- Antigen synonyms:CAMKB|CAM2|CAMK2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: CAMK2B
Clonality: Monoclonal
Clone: 2D7-D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human