Anti-ZC2HC1A Mouse Monoclonal Antibody [clone: OTI4H3]
Catalog # ORIGTA505175
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:zinc finger, C2HC-type containing 1A
- Antigen symbol:ZC2HC1A
- Clonality:Monoclonal
- Clone:OTI4H3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:34.9 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: ZC2HC1A (zinc finger, C2HC-type containing 1A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human