Anti-ERCC1 Mouse Monoclonal Antibody [clone: OTI5E6]
Catalog # ORIGTA504347
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Excision repair cross-complementation group 1
- Antigen symbol:ERCC1
- Clonality:Monoclonal
- Clone:OTI5E6
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:32.4 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
- Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: ERCC1 (excision repair cross-complementation group 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human