Anti-LDLRAP1 Mouse Monoclonal Antibody [clone: OTI7E11]
Catalog # ORIGTA503854
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Low density lipoprotein receptor adaptor protein 1
- Antigen symbol:LDLRAP1
- Clonality:Monoclonal
- Clone:OTI7E11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:33.7 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
- Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: LDLRAP1 (low density lipoprotein receptor adaptor protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human