Anti-CDCP1 Mouse Monoclonal Antibody [clone: OTI2G2]
Catalog # ORIGTA502181
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:CUB domain containing Protein 1
- Antigen symbol:CDCP1
- Clonality:Monoclonal
- Clone:OTI2G2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:92.8 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
- Recommended Dilutions: Western Blot: 1:200-500; Immunofluorescence: 1:100
Type: Primary
Antigen: CDCP1 (CUB domain containing protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human