Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-NHEJ1 Mouse Monoclonal Antibody [clone: OTI3D6]
  ORIGTA502007
 :  OriGene
undefined
Anti-NHEJ1 Mouse Monoclonal Antibody [clone: OTI3D6]
  ORIGTA502007
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    Nonhomologous end-joining factor 1
  • Antigen symbol:
    NHEJ1
  • Clonality:
    Monoclonal
  • Clone:
    OTI3D6
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    33.2 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

 

 

Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NHEJ1 (nonhomologous end-joining factor 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: