Anti-PEX5 Mouse Monoclonal Antibody [clone: OTI1D3]
Catalog # ORIGTA501389
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:peroxisomal biogenesis factor 5
- Antigen symbol:PEX5
- Clonality:Monoclonal
- Clone:OTI1D3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:69.7 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: PEX5 (peroxisomal biogenesis factor 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: