Anti-HSD17B10 Mouse Monoclonal Antibody [clone: OTI11A2]
Catalog # ORIGTA500724
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:hydroxysteroid (17-beta) dehydrogenase 10
- Antigen symbol:HSD17B10
- Clonality:Monoclonal
- Clone:OTI11A2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:26.9 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
- Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100
Type: Primary
Antigen: HSD17B10 (hydroxysteroid (17-beta) dehydrogenase 10)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human