Anti-MAPK1 Mouse Monoclonal Antibody [clone: OTI6E5]
ORIGTA500472
: OriGene
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 1
- Antigen symbol:MAPK1
- Clonality:Monoclonal
- Clone:OTI6E5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:41.4 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
- Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2-4μg/mg
Type: Primary
Antigen: MAPK1 (mitogen-activated protein kinase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: