Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-STK11 Rabbit Polyclonal Antibody
Anti-STK11 Rabbit Polyclonal Antibody
Catalog # ORIGTA317798
Supplier:  OriGene
Anti-STK11 Rabbit Polyclonal Antibody
Catalog # ORIGTA317798
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Serine/threonine Kinase 11
  • Antigen symbol:
    STK11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS, 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Pk:
    50 µG

Specifications

About this item

Recommended Dilutions: Immunochemistry: 5 µg/ml

Type: Primary
Antigen: STK11 (serine/threonine kinase 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse