Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-BAGE4 Rabbit Polyclonal Antibody
Anti-BAGE4 Rabbit Polyclonal Antibody
Catalog # ORIGTA315954
Supplier:  OriGene
Anti-BAGE4 Rabbit Polyclonal Antibody
Catalog # ORIGTA315954
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    B melanoma antigen family, member 4
  • Antigen symbol:
    BAGE4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: BAGE4 (B melanoma antigen family, member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human