Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: AC-16]
Catalog # ORIGTA309389
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Heat shock protein 90kDa alpha (cytosolic), class B member 1
- Antigen symbol:HSP90AB1
- Clonality:Monoclonal
- Clone:AC-16
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS with 50% glycerol and 0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
- Recommended Dilutions:
Type: Primary
Antigen: HSP90AB1 (heat shock protein 90kDa alpha (cytosolic), class B member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: