Mouse IgG2b Isotype Control [clone: MPC-11] (violetFluor® 450)
Catalog # 75-4732-U100
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:MPC-11
- Conjugation:VioletFluor® 450
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.
Type: Control
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: monoclonal
Clone: MPC-11
Conjugation: violetFluor® 450
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: