Order Entry
Northern Ireland
ContactUsLinkComponent
Mouse IgG2b Isotype Control [clone: MPC-11] (FITC (Fluorescein Isothiocyanate))
Mouse IgG2b Isotype Control [clone: MPC-11] (FITC (Fluorescein Isothiocyanate))
Catalog # 35-4732-U100
Supplier:  Tonbo Biosciences
Mouse IgG2b Isotype Control [clone: MPC-11] (FITC (Fluorescein Isothiocyanate))
Catalog # 35-4732-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    MPC-11
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.

Type: Control
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: monoclonal
Clone: MPC-11
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: