Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-CD27 Mouse Monoclonal Antibody (APC (Allophycocyanin))/Cy7®) [clone: O323]
 
undefined
Anti-CD27 Mouse Monoclonal Antibody (APC (Allophycocyanin))/Cy7®) [clone: O323]

 

  • Catalog No:
  • 25-0279-T100
  • 25-0279-T025
  • Antigen symbol:
  • CD27
  • CD27
  • Antigen name:
  • CD27 molecule
  • CD27 molecule
  • Antigen synonyms:
  • Tp55|S152|T14|TNFRSF7|CD27|S152. LPFS2
  • Tp55|S152|T14|TNFRSF7|CD27|S152. LPFS2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)/Cy7®
  • APC (Allophycocyanin)/Cy7®
  • Clone:
  • O323
  • O323
  • Reactivity:
  • Baboon, Cynomolgus monkey, Macaque, Human
  • Baboon, Cynomolgus monkey, Macaque, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 939
  • 939
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

Frequently Bought Together