Anti-Protein Kinase D2 (PKC-D2) Rabbit Polyclonal Antibody
Catalog # ABGEAP7934A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Protein kinase D2
- Antigen symbol:PRKD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal (952–968 aa)
- Cross adsorption:No
- Gene ID:2494
- Antigen synonyms:B1F|B1F2|FTZ-F1beta|LRH-1|FTZ-F1|hB1F-2|LRH1|FTF|CPF
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:EQANRSKHEKLSTFC
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Immunogen:This NR5A2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 314-342 amino acids from the Central region of human NR5A2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: Protein Kinase D2 (PKC-D2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human