Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-GRIN2A Rabbit Polyclonal Antibody [clone: 220ct16.4.3.1]
Anti-GRIN2A Rabbit Polyclonal Antibody [clone: 220ct16.4.3.1]
Catalog # ABGEAP11390B
Supplier:  Abgent
Anti-GRIN2A Rabbit Polyclonal Antibody [clone: 220ct16.4.3.1]
Catalog # ABGEAP11390B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate Ionotropic Receptor NMDA Type Subunit 2A
  • Antigen symbol:
    GRIN2A
  • Clonality:
    Polyclonal
  • Clone:
    220ct16.4.3.1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Antigen synonyms:
    NR2A|LKS|glutamate ionotropic receptor NMDA type subunit 2A|EPND|GluN2A|NMDAR2A|FESD
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: GRIN2A (glutamate receptor, ionotropic, N-methyl D-aspartate 2A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human