Anti-Nitrotyrosine Rabbit Polyclonal Antibody
Catalog # BOSSBS-8551R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Nitrotyrosine
- Antigen symbol:Nitro-Tyr
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Multiple
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Nitrotyrosine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others