Peqlab, PCR Tubes
Catalog # 732-3215
Supplier: VWR Peqlab
Specifications
- Packed:2 bags of 500
- Capacity:0,2 ml
- Colour:Clear
- Description:PCR tubes with flat cap
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:IRECS
- Pk:1.000
Specifications
About this item
These PCR tubes with caps are made of PP resin and are designed to fit most popular brands of thermal cyclers.
- Ultra-thin walls for rapid heat transfer
- Certified free from DNase, RNase and human DNA
- Manufactured from polypropylene under strict quality control guidelines
Available with domed or flat caps.