Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), EMPROVE® EXPERT BP, Ph. Eur., ACS, JP, USP, high purity for biopharmaceutical production
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
CAS Number:77-86-1
:
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679
Boiling Pt:357
Melting Pt:169…172 °C
Density:1.328 g/cm³ (20 °C)
Molecular Weight:121.14 g/mol
Merck Index:13,09842
Storage Temperature:Ambient
EINECS:201-064-4
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together