ContactUsLinkComponent
Safety waste systems, SP Bel-Art
Safety waste systems, SP Bel-Art
Catalog # 212-0731
undefined
Safety waste systems, SP Bel-Art
Catalog # 212-0731
Specifications
  • Ø opening:
    64 mm
  • Capacity:
    20 L
  • Colour:
    Translucent
  • Height:
    381 mm
  • Pk:
    1
Specifications
About this item
HDPE, translucent, graduated.
  • Translucent for observing content level
  • Includes screw-top to seal container for safe transit
  • Flared and angled funnel for easy filling
A system for safe handling and temporary storage of spent solvents or other liquid waste. Features a screw-on funnel top with snap-on cap seals to retain fumes.