Burners
Supplier: R L ENTERPRISES
Certificates
About this item
Burners are constructed using a heavy pressure diecast base painted in a blue acrylic which is chemically resistant. It is suitable for natural gas.
- All burners are tested for flame quality and leakage prior to packing
- The tests are carried out at the standard gas pressure for each specific type gas
The hottest flame temperature for all natural gas burners is between 900 and 1000 °C. The temperature is achieved at the tip of the blue flame cone. Typically the temperature of the airless yellow flame will be between 750 and 800 °C.
Frequently Bought Together