ContactUsLinkComponent
Brilliant green bile broth, tubes
  HACH32215
 :  Hach
 :  
undefined
Brilliant green bile broth, tubes
  HACH32215
 :  Hach
 :  
  • Pk:
    15
  • Standards:
    -
  • Description:
    Brilliant green broth tubes, glass
  • Volume:
    15 ml
They are used in the applications of drinking water, wastewater, ambient water, food and beverage. And it is a complete solution for testing drinking water also.
  • Saves time
  • Saves labor cost for ready to use MPN tubes