Order Entry
Northern Ireland
ContactUsLinkComponent
 
VWR® ARISTAR®, ICP Multi-Element Solution According USP232 Parental dose 6 Elements 100 mg/L in 15% Hydrochloric Acid
Catalog # 85038.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, ICP Multi-Element Solution According USP232 Parental dose 6 Elements 100 mg/L in 15% Hydrochloric Acid
Catalog # 85038.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    ICP multi element solution according USP 232 Parental dose 6 elements, 100 mg/L

Specifications

About this item

Six elements: 100 mg/L each of Ir; Pt; Os; Rh; Pd; Ru in HCl 15%.

Certifications: Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.