NFKB p65 (rel A) (phospho-Ser276) control peptide
Catalog # ROCK000-000-264
Supplier: Rockland Immunochemicals
CAS Number:
Specifications
- Pk:50 µG
- Seq:01
- Protein/peptide name:NFKB p65 (rel A) (phospho-Ser276) Control Peptide
- Tested applications:Control Peptide