Order Entry
Northern Ireland
ContactUsLinkComponent
Sodium ammonium hydrogen phosphate tetrahydrate, EMSURE® for analysis, Supelco®
Sodium ammonium hydrogen phosphate tetrahydrate, EMSURE® for analysis, Supelco®
Catalog # 1.06682.1000
Supplier:  Merck
CAS Number:  
Sodium ammonium hydrogen phosphate tetrahydrate, EMSURE® for analysis, Supelco®
Catalog # 1.06682.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Solubility:
    167 g/l (20 °C)
  • Melting Point:
    79 °C
  • Molar Mass:
    209.07 g/mol
  • Density:
    0.85 g/cm3 (20 °C)
  • pH:
    8.3 (50 g/l, H2O, 20 °C)

Specification Test Results

About this item

CAS Number:7783-13-3
 :
Formula:NaNH₄HPO₄·4 H₂O
 :209.07 g/mol
 :M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:235-860-8
 :MFCD00066293
Formula:NaNH₄HPO₄·4H₂O
MDL Number:MFCD00066970