, HiPerSolv CHROMANORM® for LC-MS
:75-05-8
:3 II
:Packaged under nitrogen.
:200-835-2
:1648
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:
- Nominal concentration (V/V):0.095 - 0.105 %
- Evaporation residue:Max. 0.0001 %
- Water:Max. 0.005 %
- Ca (Calcium):Max. 0.2 ppm
- K (Potassium):Max. 0.2 ppm
- Mg (Magnesium):Max. 0.2 ppm
- Na (Sodium):Max. 0.5 ppm
- Transmittance (235 nm):Min. 20 %
- Transmittance (245 nm):Min. 50 %
- Transmittance (250 nm):Min. 80 %
- Transmittance (260 nm):Min. 95 %
- Transmittance (270 nm):Min. 99 %
- Suitable for LC-MS:Passes test
Frequently Bought Together