Order Entry
Northern Ireland
ContactUsLinkComponent
Tetrahydrofuran : Acetic anhydride (87.5:12.5) (v/v)
Tetrahydrofuran : Acetic anhydride (87.5:12.5) (v/v)
Catalog # VWRS83929.410
Supplier:  VWR Collection
CAS Number:  
Tetrahydrofuran : Acetic anhydride (87.5:12.5) (v/v)
Catalog # VWRS83929.410
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    10 L
  • Seq:
    5214

Specifications

About this item

 :
Density:0,9060 g/cm³ (20 °C)