Order Entry
Northern Ireland
ContactUsLinkComponent
Benzyl alcohol ≥99.5%, BAKER ANALYZED®, J.T.Baker®
Irritant
Benzyl alcohol ≥99.5%, BAKER ANALYZED®, J.T.Baker®
Catalog # 9050-01
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Benzyl alcohol ≥99.5%, BAKER ANALYZED®, J.T.Baker®
Catalog # 9050-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.5 %
  • Assay (C₆H₅CH₂OH) (by GC, corrected for water):
    ≥99.5 %
  • Color (APHA):
    ≤10
  • Residue after Ignition:
    ≤0.005 %
  • Solubility in H₂O:
    Passes Test
  • Chlorinated Compounds (as Cl):
    Passes Test
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.05 %

Specification Test Results

About this item

 :
 :108.14 g/mol
Merck Index:13,01126
Boiling Pt:205 °C (1013 hPa)
Melting Pt:–15,3 °C
Density:1,05 g/cm³ (20 °C)
Flash Pt:101 °C
Storage Temperature:Ambient
EINECS:202-859-9
 :GHS07
CAS Number:100-51-6
Formula:C₆H₅CH₂OH
MDL Number:MFCD00004599