Order Entry
Northern Ireland
ContactUsLinkComponent
Phenol red sodium salt, powder, BAKER ANALYZED® ACS, J.T.Baker®
Phenol red sodium salt, powder, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # T265-01
Supplier:  Avantor
CAS Number:  
undefined
Phenol red sodium salt, powder, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # T265-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, wide mouth, black T/E cap
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    Passes Test
  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • ACS - Clarity of Solution:
    Passes Test
  • Visual Transition Interval - pH…8.2 (Red):
    Passes Test
  • Visual Transition Interval - pH…6.8 (Yellow):
    Passes Test

Specification Test Results

About this item

 :
 :376.35 g/mol
Merck Index:12,07397
Melting Pt:285 °C
Storage Temperature:Ambient
EINECS:252-057-8
CAS Number:34487-61-1
Formula:C₁₉H₁₃NaO₅S
MDL Number:MFCD00066901