Order Entry
Northern Ireland
ContactUsLinkComponent
PCR clean-up, PCR normalizer kit, Axygen® AxyPrep™ Mag
PCR clean-up, PCR normalizer kit, Axygen® AxyPrep™ Mag
Catalog # MAG-PCR-NM-5
Supplier:  Corning
CAS Number:  
undefined
PCR clean-up, PCR normalizer kit, Axygen® AxyPrep™ Mag
Catalog # MAG-PCR-NM-5
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    Axygen® AxyPrep™ Mag PCR Normalizer kit, 96 Preps
  • Description:
    Axygen® AxyPrep™ Mag PCR Normalizer Kit
  • Volume:
    5 ml

Specifications

About this item

The AxyPrep™ Mag PCR Normalizer Kit utilizes a paramagnetic bead-based purification system for PCR clean up.

  • Dual action: Normalization and post PCR clean-up enables significant reduction in processing and faster turnaround times
  • Scalable: Tube, 96-, and 384-well plate formats
  • Manual and automation-compatible protocol
  • Compatible with several downstream applications

Axygen® AxyPrep™ Mag PCR Normalizer Kit utilises a paramagnetic bead-based purification system for PCR clean up. AxyPrep™ Mag PCR Normalizer beads have limited binding surface, therefore by limiting the amount of beads added in a given purification reaction, pre-defined amounts of DNA can be isolated based on user need. PCR normalisation is accomplished during this purification process so that additional DNA quantification and dilution are not necessary.

Time, labour and reagent cost are greatly saved with the AxyPrep™ Mag normalisation purification chemistry and thus efficiently streamlining genomics downstream applications. The protocol mainly consists of binding, washing and elution steps, and can be performed directly in the thermal cycling plate and requires no centrifugation or filtration. The process can be automated for high throughput applications.