Double bossheads
Catalog # 241-0156
Supplier: R L ENTERPRISES
Specifications
- Material:Metal/plastic
- Pk:1
- For rod Ø:16 mm
Specifications
About this item
Double bosshead made from cast alloy.
- With blue plastic headed nickel plated screws
- For use with rods up to 16 mm