Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
SATA (N-Succinimidyl S-Acetylthioglycolate)
SATA (N-Succinimidyl S-Acetylthioglycolate)
SATA (N-Succinimidyl S-Acetylthioglycolate)
SATA (N-Succinimidyl S-Acetylthioglycolate)
Catalog # BC96
Supplier:
G-Biosciences
CAS Number:
Expand image
SATA (N-Succinimidyl S-Acetylthioglycolate)
Catalog # BC96
Supplier:
G-Biosciences
CAS Number:
Specifications
Pk:
100 mg
Seq:
2440
Specifications
About this item
Product Details
:
:
231.22 g/mol
CAS Number:
76931-93-6
Formula:
C₈H₉NO₅S
MDL Number:
MFCD00036891
Product Details